Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(4-Nitrophenyl)furfural, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
7147-77-5
C11H7NO4
217.18
RTSOJVJDKNKNFU-UHFFFAOYSA-N
5-4-nitrophenyl-2-furaldehyde,5-4-nitrophenyl furan-2-carbaldehyde,5-4-nitrophenyl-2-furancarboxaldehyde,5-4-nitrophenyl furfural,5-4-nitro-phenyl-furan-2-carbaldehyde,2-furancarboxaldehyde, 5-4-nitrophenyl,5-p-nitrophenyl furfural,5-4-nitrophenyl 2-furaldehyde,cambridge id 5143178,acmc-1bl92
81567
5-(4-nitrophenyl)furan-2-carbaldehyde
C1=CC(=CC=C1C2=CC=C(O2)C=O)[N+](=O)[O-]
7147-77-5
C11H7NO4
217.18
RTSOJVJDKNKNFU-UHFFFAOYSA-N
5-4-nitrophenyl-2-furaldehyde,5-4-nitrophenyl furan-2-carbaldehyde,5-4-nitrophenyl-2-furancarboxaldehyde,5-4-nitrophenyl furfural,5-4-nitro-phenyl-furan-2-carbaldehyde,2-furancarboxaldehyde, 5-4-nitrophenyl,5-p-nitrophenyl furfural,5-4-nitrophenyl 2-furaldehyde,cambridge id 5143178,acmc-1bl92
81567
5-(4-nitrophenyl)furan-2-carbaldehyde
C1=CC(=CC=C1C2=CC=C(O2)C=O)[N+](=O)[O-]
Melting Point 204°C to 208°C
Quantity 1g
Packaging Glass bottle
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 217.18
Percent Purity 98%
Chemical Name or Material 5-(4-Nitrophenyl)furfural

GHS Signal Word: Warning

EINECSNumber : 230-459-4