Alfa Aesar™ 5,5'-Dibromo-BAPTA, 95%

Manufacturer: Alfa Aesar™ A11388-06

Quantity 5g

 View more versions of this product

Catalog No. AAA1138806

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 73630-11-2
    Molecular Formula C22H22Br2N2O10
    Formula Weight 634.23
    MDL Number MFCD00036169
    Synonym 1,2-Bis(2-amino-5-bromophenoxy)ethane-N,N,N',N'-tetraacetic acid
    Chemical Name or Material 5,5'-Dibromo-BAPTA

    Specifications

    Formula Weight 634.23
    CAS 73630-11-2
    Molecular Formula C22H22Br2N2O10
    Synonym 1,2-Bis(2-amino-5-bromophenoxy)ethane-N,N,N',N'-tetraacetic acid
    Recommended Storage 2°C to 8°C
    Melting Point 172° to 178°C
    Quantity 5g
    MDL Number MFCD00036169
    Chemical Name or Material 5,5'-Dibromo-BAPTA
    Assay 95%

    2°C to 8°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit