Alfa Aesar™ 5,5-Dimethyl-1,3-dioxane-2-butanal, 96%

Manufacturer: Alfa Aesar™ B2185809

Quantity 10g

 View more versions of this product

Catalog No. AAB2185809

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 127600-13-9
    Molecular Formula C10H18O3
    Formula Weight 186.25
    MDL Number MFCD02683076
    Synonym 5,5-Dimethyl-2-(3-formylpropyl)-1,3-dioxane; Glutaraldehyde mono-neopentyl glycol acetal
    Chemical Name or Material 5,5-Dimethyl-1,3-dioxane-2-butanal

    Specifications

    Formula Weight 186.25
    Molecular Formula C10H18O3
    MDL Number MFCD02683076
    CAS 127600-13-9
    Quantity 10g
    Synonym 5,5-Dimethyl-2-(3-formylpropyl)-1,3-dioxane; Glutaraldehyde mono-neopentyl glycol acetal
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 5,5-Dimethyl-1,3-dioxane-2-butanal
    Assay 96%

    Description

    Applications
    It is employed as a intermediate for pharmaceuticals.

    Notes
    Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit