Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5-Dimethylhydantoin, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
250g
1kg
5kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
77-71-4
C5H8N2O2
128.131
MFCD00005266
YIROYDNZEPTFOL-UHFFFAOYSA-N
5,5-dimethylhydantoin,dimethylhydantoin,dm hydantoin,dantoin dmh,2,4-imidazolidinedione, 5,5-dimethyl,5,5-dimethyl-2,4-imidazolidinedione,dantoin 736,hydantoin, 5,5-dimethyl,5,5-dimethylhydantoin dmh,5,5-dimethyl hydantoin
6491
5,5-dimethylimidazolidine-2,4-dione
CC1(C(=O)NC(=O)N1)C
77-71-4
C5H8N2O2
128.131
MFCD00005266
YIROYDNZEPTFOL-UHFFFAOYSA-N
5,5-dimethylhydantoin,dimethylhydantoin,dm hydantoin,dantoin dmh,2,4-imidazolidinedione, 5,5-dimethyl,5,5-dimethyl-2,4-imidazolidinedione,dantoin 736,hydantoin, 5,5-dimethyl,5,5-dimethylhydantoin dmh,5,5-dimethyl hydantoin
6491
5,5-dimethylimidazolidine-2,4-dione
CC1(C(=O)NC(=O)N1)C
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Melting Point 175°C to 180°C
Loss on Drying 0.25% max. (60°C, 2 hrs, 1mm Hg)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 24, 289
Fieser 07,126
Formula Weight 128.13
Physical Form Crystalline Powder or Crystals
Percent Purity ≥96.0%
Chemical Name or Material 5, 5-Dimethylhydantoin
Show More Show Less