Alfa Aesar™ 5,5-Dimethyloxazolidine-2,4-dione, 99%

Manufacturer: Alfa Aesar™ A1071406

Quantity 5g

 View more versions of this product

Catalog No. AAA1071406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 695-53-4
    Molecular Formula C5H7NO3
    Formula Weight 129.12
    MDL Number MFCD00005379
    Beilstein 113541
    Merck Index 14,3213
    Synonym Dimethadione
    Chemical Name or Material 5,5-Dimethyloxazolidine-2,4-dione

    Specifications

    Formula Weight 129.12
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Synonym Dimethadione
    RTECS Number RP9100000
    Molecular Formula C5H7NO3
    Melting Point 77° to 80°C
    MDL Number MFCD00005379
    Quantity 5g
    Merck Index 14,3213
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H361
    Suspected of damaging fertility or the unborn child.
    CAS 695-53-4
    Beilstein 113541
    Chemical Name or Material 5,5-Dimethyloxazolidine-2,4-dione
    Assay 99%

    GHS H Statement
    H361
    Suspected of damaging fertility or the unborn child.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit