Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5-Dimethyloxazolidine-2,4-dione, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
695-53-4
C5H7NO3
129.115
MFCD00005379
JYJFNDQBESEHJQ-UHFFFAOYSA-N
dimethadione,5,5-dimethyloxazolidine-2,4-dione,5,5-dimethyl-2,4-oxazolidinedione,propazone,dimethadion,eupractone,dimethyloxazolidinedione,2,4-oxazolidinedione, 5,5-dimethyl,5,5-dimethyloxazolidinedione,dimetadione dcit
3081
5,5-dimethyl-1,3-oxazolidine-2,4-dione
CC1(C(=O)NC(=O)O1)C
695-53-4
C5H7NO3
129.115
MFCD00005379
JYJFNDQBESEHJQ-UHFFFAOYSA-N
dimethadione,5,5-dimethyloxazolidine-2,4-dione,5,5-dimethyl-2,4-oxazolidinedione,propazone,dimethadion,eupractone,dimethyloxazolidinedione,2,4-oxazolidinedione, 5,5-dimethyl,5,5-dimethyloxazolidinedione,dimetadione dcit
3081
5,5-dimethyl-1,3-oxazolidine-2,4-dione
CC1(C(=O)NC(=O)O1)C
Appearance (5% aq. soln.) clear, colorless
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 75.0°C to 77.0°C
Color White
Quantity 1g
Boiling Point 137.0°C (6.0 mmHg)
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 27, 252
Merck Index 15, 3235
Formula Weight 129.12
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 5, 5-Dimethyloxazolidine-2, 4-dione, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause damage to organs through prolonged or repeated exposure.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 211-781-4

RTECSNumber : RP9100000

TSCA : TSCA