Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5-Diphenylhydantoin, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
Plastic bottle
57-41-0
C15H12N2O2
252.27
MFCD00005264
CXOFVDLJLONNDW-UHFFFAOYSA-N
phenytoin,5,5-diphenylhydantoin,diphenylhydantoin,dilantin,phenytoine,dihydantoin,aleviatin,dilabid,diphantoin,diphenylan
1775
CHEBI:8107
5,5-diphenylimidazolidine-2,4-dione
C1=CC=C(C=C1)C2(C(=O)NC(=O)N2)C3=CC=CC=C3
57-41-0
C15H12N2O2
252.27
MFCD00005264
CXOFVDLJLONNDW-UHFFFAOYSA-N
phenytoin,5,5-diphenylhydantoin,diphenylhydantoin,dilantin,phenytoine,dihydantoin,aleviatin,dilabid,diphantoin,diphenylan
1775
CHEBI:8107
5,5-diphenylimidazolidine-2,4-dione
C1=CC=C(C=C1)C2(C(=O)NC(=O)N2)C3=CC=CC=C3
Melting Point 293.5°C to 295°C
Packaging Plastic bottle
Loss on Drying 0.5% max. (1 g, 105°C)
Infrared Spectrum Authentic
Color White
Quantity 500g
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Beilstein 24, 410
Merck Index 15, 7433
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alkali hydroxides,1g/60mL alcohol,1g/30mL acetone
Formula Weight 252.27
Physical Form Powder
Percent Purity 99+%
Chemical Name or Material 5, 5-Diphenylhydantoin, 99+%
Show More Show Less

GHS H Statement:
Suspected of causing cancer.
Harmful if swallowed.
Suspected of damaging the unborn child.

GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-328-6

RTECSNumber : MU1050000