Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6,7,8,9,10-Hexahydrocyclohept[b]indole, 98%, Alfa Aesar™

Click to view available options
Quantity:
1g
5g
25g
2047-89-4
C13H15N
185.27
MFCD00101340
XZUJMYLNFZHNLP-UHFFFAOYSA-N
5,6,7,8,9,10-hexahydrocyclohepta b indole,5,6,7,8,9,10-hexahydro-cyclohepta b indole,5h,6h,7h,8h,9h,10h-cyclohepta b indole,cycloheptan a indole,2,3-pentano-1h-indole,5,6,7,8,9,10-hexahydrocyclohept b indole,5,6,7,8,9,10-hexahydrocyclohepta b indole #,6,7,8,9,10,10a-hexahydrocyclohepta b indole,6,7,8,9,10-pentahydrocyclohepta 1,2-b indole,6,7,8,9,10-pentahydrocyclohepta 2,1-b indole
251955
5,6,7,8,9,10-hexahydrocyclohepta[b]indole
C1CCC2=C(CC1)NC3=CC=CC=C23
2047-89-4
C13H15N
185.27
MFCD00101340
XZUJMYLNFZHNLP-UHFFFAOYSA-N
5,6,7,8,9,10-hexahydrocyclohepta b indole,5,6,7,8,9,10-hexahydro-cyclohepta b indole,5h,6h,7h,8h,9h,10h-cyclohepta b indole,cycloheptan a indole,2,3-pentano-1h-indole,5,6,7,8,9,10-hexahydrocyclohept b indole,5,6,7,8,9,10-hexahydrocyclohepta b indole #,6,7,8,9,10,10a-hexahydrocyclohepta b indole,6,7,8,9,10-pentahydrocyclohepta 1,2-b indole,6,7,8,9,10-pentahydrocyclohepta 2,1-b indole
251955
5,6,7,8,9,10-hexahydrocyclohepta[b]indole
C1CCC2=C(CC1)NC3=CC=CC=C23
Quantity 25g
Melting Point 143°C to 146°C
Formula Weight 185.27
Percent Purity 98%
Chemical Name or Material 5,6,7,8,9,10-Hexahydrocyclohept[b]indole

TSCA : No

Recommended Storage : Ambient temperatures