Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6,7,8-Tetrahydro-2-naphthol, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
1125-78-6
C10H12O
148.2
MFCD00001738
UMKXSOXZAXIOPJ-UHFFFAOYSA-N
5,6,7,8-tetrahydro-2-naphthol,6-tetralinol,6-hydroxytetralin,2-naphthalenol, 5,6,7,8-tetrahydro,5,6,7,8-tetrahydro-beta-naphthol,ac-beta-tetralol,5,6,7,8-tetrahydro-2-hydroxynaphthalene,2-naphthol, 5,6,7,8-tetrahydro,unii-tmr02i7n8s,5,6,7,8-tetrahydro-2-naphthalenol
14305
CHEBI:34448
5,6,7,8-tetrahydronaphthalen-2-ol
C1CCC2=C(C1)C=CC(=C2)O
1125-78-6
C10H12O
148.2
MFCD00001738
UMKXSOXZAXIOPJ-UHFFFAOYSA-N
5,6,7,8-tetrahydro-2-naphthol,6-tetralinol,6-hydroxytetralin,2-naphthalenol, 5,6,7,8-tetrahydro,5,6,7,8-tetrahydro-beta-naphthol,ac-beta-tetralol,5,6,7,8-tetrahydro-2-hydroxynaphthalene,2-naphthol, 5,6,7,8-tetrahydro,unii-tmr02i7n8s,5,6,7,8-tetrahydro-2-naphthalenol
14305
CHEBI:34448
5,6,7,8-tetrahydronaphthalen-2-ol
C1CCC2=C(C1)C=CC(=C2)O
Infrared Spectrum Authentic
Boiling Point 275°C to 276°C
Flash Point >112°C
Color Beige
Packaging Glass bottle
Melting Point 58°C to 62°C
Quantity 10g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Beilstein 06, 579
Formula Weight 148.2
Physical Form Crystalling Mass
Percent Purity 98%
Chemical Name or Material 5,6,7,8-Tetrahydro-2-naphthol, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 214-410-4

RTECSNumber : QL5300000