Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6,7,8-Tetrahydro-2-naphthol 98.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
1125-78-6
C10H12O
148.205
MFCD00001738
UMKXSOXZAXIOPJ-UHFFFAOYSA-N
5,6,7,8-tetrahydro-2-naphthol,6-tetralinol,6-hydroxytetralin,2-naphthalenol, 5,6,7,8-tetrahydro,5,6,7,8-tetrahydro-beta-naphthol,ac-beta-tetralol,5,6,7,8-tetrahydro-2-hydroxynaphthalene,2-naphthol, 5,6,7,8-tetrahydro,unii-tmr02i7n8s,5,6,7,8-tetrahydro-2-naphthalenol
14305
CHEBI:34448
5,6,7,8-tetrahydronaphthalen-2-ol
C1CCC2=C(C1)C=CC(=C2)O
1125-78-6
C10H12O
148.205
MFCD00001738
UMKXSOXZAXIOPJ-UHFFFAOYSA-N
5,6,7,8-tetrahydro-2-naphthol,6-tetralinol,6-hydroxytetralin,2-naphthalenol, 5,6,7,8-tetrahydro,5,6,7,8-tetrahydro-beta-naphthol,ac-beta-tetralol,5,6,7,8-tetrahydro-2-hydroxynaphthalene,2-naphthol, 5,6,7,8-tetrahydro,unii-tmr02i7n8s,5,6,7,8-tetrahydro-2-naphthalenol
14305
CHEBI:34448
5,6,7,8-tetrahydronaphthalen-2-ol
C1CCC2=C(C1)C=CC(=C2)O
Color Undesignated
Boiling Point 133°C
Quantity 500g
Melting Point 62°C
Formula Weight 148.21
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 5,6,7,8-Tetrahydro-2-naphthol

RTECSNumber : QL5300000

TSCA : No