Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6,7,8-Tetrahydroisoquinoline, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
36556-06-6
C9H11N
133.19
HTMGQIXFZMZZKD-UHFFFAOYSA-N
5,6,7,8-tetrahydro-isoquinoline,isoquinoline, 5,6,7,8-tetrahydro,acmc-1ct7z,5,6,7,8-tetra-hydro-isoquinoline,5,6,7,8-tetrahydroisoquinoline
119010
5,6,7,8-tetrahydroisoquinoline
C1CCC2=C(C1)C=CN=C2
36556-06-6
C9H11N
133.19
HTMGQIXFZMZZKD-UHFFFAOYSA-N
5,6,7,8-tetrahydro-isoquinoline,isoquinoline, 5,6,7,8-tetrahydro,acmc-1ct7z,5,6,7,8-tetra-hydro-isoquinoline,5,6,7,8-tetrahydroisoquinoline
119010
5,6,7,8-tetrahydroisoquinoline
C1CCC2=C(C1)C=CN=C2
Quantity 25g
Color Yellow
Boiling Point 77.0°C to 81.0°C (1.0 Torr)
Flash Point 100°C
Density 1.0400g/mL
Packaging Glass bottle
Specific Gravity 1.04
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Formula Weight 133.19
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 5,6,7,8-Tetrahydroisoquinoline
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 253-098-4