Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5,6,7-Trimethoxyflavone, 97%

Click to view available options
Quantity:
0.25g
1g
973-67-1
C18H16O5
312.321
MFCD00017457
HJNJAUYFFFOFBW-UHFFFAOYSA-N
5,6,7-trimethoxyflavone,baicalein trimethyl ether,baicalein 5,6,7-trimethyl ether,baicalein-5,6,7-trimethylether,5,6,7-trimethoxy-2-phenyl-4h-chromen-4-one,5,6,7-trimethoxy-2-phenyl-chromen-4-one,baicalein-5,6,7-trimethyl ether,pubchem9835,5,6,7-trimethylbaicalein
442583
CHEBI:2980
5,6,7-trimethoxy-2-phenylchromen-4-one
COC1=C(C(=C2C(=C1)OC(=CC2=O)C3=CC=CC=C3)OC)OC
973-67-1
C18H16O5
312.321
MFCD00017457
HJNJAUYFFFOFBW-UHFFFAOYSA-N
5,6,7-trimethoxyflavone,baicalein trimethyl ether,baicalein 5,6,7-trimethyl ether,baicalein-5,6,7-trimethylether,5,6,7-trimethoxy-2-phenyl-4h-chromen-4-one,5,6,7-trimethoxy-2-phenyl-chromen-4-one,baicalein-5,6,7-trimethyl ether,pubchem9835,5,6,7-trimethylbaicalein
442583
CHEBI:2980
5,6,7-trimethoxy-2-phenylchromen-4-one
COC1=C(C(=C2C(=C1)OC(=CC2=O)C3=CC=CC=C3)OC)OC
Odor Odorless
Melting Point 165°C to 167°C
Quantity 0.25g
Formula Weight 312.33
Percent Purity 97%
Chemical Name or Material 5,6,7-Trimethoxyflavone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning