Learn More
Aat BioquestSupplier Diversity Partner 5(6)-CR6G [5-(and 6)-Carboxyrhodamine 6G] DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 330
In general, rhodamine 6G conjugates have higher fluorescence quantum yields than tetramethylrhodamine conjugates, as well as good photostability. 5(6)-CR6G is the mixture of two carboxy rhodamine 6G isomers.
Catalog No. 50-195-2835