Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5,6-Dichloronicotinic acid, 98+%

Click to view available options
Quantity:
5g
25g
41667-95-2
C6H3Cl2NO2
191.995
MFCD00075181
RNRLTTNKVLFZJS-UHFFFAOYSA-N
5,6-dichloronicotinic acid,5,6-dichloro nicotinic acid,5,6-dichloro-3-pyridinecarboxylic acid,5,6-dichloronicotinicacid,2,3-dichloro-5-carboxypyridine,3-pyridinecarboxylic acid, 5,6-dichloro,5,6-dichloro-nicotinic acid,pubchem12968,acmc-209jl4,5,6 dichloronicotinic acid
247987
5,6-dichloropyridine-3-carboxylic acid
C1=C(C=NC(=C1Cl)Cl)C(=O)O
41667-95-2
C6H3Cl2NO2
191.995
MFCD00075181
RNRLTTNKVLFZJS-UHFFFAOYSA-N
5,6-dichloronicotinic acid,5,6-dichloro nicotinic acid,5,6-dichloro-3-pyridinecarboxylic acid,5,6-dichloronicotinicacid,2,3-dichloro-5-carboxypyridine,3-pyridinecarboxylic acid, 5,6-dichloro,5,6-dichloro-nicotinic acid,pubchem12968,acmc-209jl4,5,6 dichloronicotinic acid
247987
5,6-dichloropyridine-3-carboxylic acid
C1=C(C=NC(=C1Cl)Cl)C(=O)O
Melting Point ~164°C (decomposition)
Quantity 5g
Beilstein 383740
Formula Weight 192
Percent Purity ≥98%
Chemical Name or Material 5,6-Dichloronicotinic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning