Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Dichloronicotinic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
41667-95-2
C6H3Cl2NO2
191.995
MFCD00075181
RNRLTTNKVLFZJS-UHFFFAOYSA-N
5,6-dichloronicotinic acid,5,6-dichloro nicotinic acid,5,6-dichloro-3-pyridinecarboxylic acid,5,6-dichloronicotinicacid,2,3-dichloro-5-carboxypyridine,3-pyridinecarboxylic acid, 5,6-dichloro,5,6-dichloro-nicotinic acid,pubchem12968,acmc-209jl4,5,6 dichloronicotinic acid
247987
5,6-dichloropyridine-3-carboxylic acid
C1=C(C=NC(=C1Cl)Cl)C(=O)O
41667-95-2
C6H3Cl2NO2
191.995
MFCD00075181
RNRLTTNKVLFZJS-UHFFFAOYSA-N
5,6-dichloronicotinic acid,5,6-dichloro nicotinic acid,5,6-dichloro-3-pyridinecarboxylic acid,5,6-dichloronicotinicacid,2,3-dichloro-5-carboxypyridine,3-pyridinecarboxylic acid, 5,6-dichloro,5,6-dichloro-nicotinic acid,pubchem12968,acmc-209jl4,5,6 dichloronicotinic acid
247987
5,6-dichloropyridine-3-carboxylic acid
C1=C(C=NC(=C1Cl)Cl)C(=O)O
Melting Point 163°C to 168°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color Tan to White
Water 0.5% max. (K.F.)
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 192
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 5, 6-Dichloronicotinic acid
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.