Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5,6-Dichloropyridine-2-carboxylic acid, 98%

Click to view available options
Quantity:
250mg
1g
5g
88912-24-7
C6H3Cl2NO2
191.995
MFCD13185546
QOJNAEPFSUYAFL-UHFFFAOYSA-N
5,6-dichloropicolinic acid,5,6-dichloro-2-pyridinecarboxylic acid,2-pyridinecarboxylic acid, 5,6-dichloro,2,3-dichloro-6-carboxypyridine,2-pyridinecarboxylicacid, 5,6-dichloro,acmc-209vdz,2,3-dichloro-6-pyridinecarboxylicacid,5,6-dichloro-pyridine-2-carboxylic acid
13278016
5,6-dichloropyridine-2-carboxylic acid
C1=CC(=NC(=C1Cl)Cl)C(=O)O
88912-24-7
C6H3Cl2NO2
191.995
MFCD13185546
QOJNAEPFSUYAFL-UHFFFAOYSA-N
5,6-dichloropicolinic acid,5,6-dichloro-2-pyridinecarboxylic acid,2-pyridinecarboxylic acid, 5,6-dichloro,2,3-dichloro-6-carboxypyridine,2-pyridinecarboxylicacid, 5,6-dichloro,acmc-209vdz,2,3-dichloro-6-pyridinecarboxylicacid,5,6-dichloro-pyridine-2-carboxylic acid
13278016
5,6-dichloropyridine-2-carboxylic acid
C1=CC(=NC(=C1Cl)Cl)C(=O)O
Quantity 250mg
Beilstein 4390497
Formula Weight 192
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 5,6-Dichloropyridine-2-carboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning