Alfa Aesar™ 5,6-Difluorosalicylic acid, 98%

Manufacturer: Alfa Aesar™ H31634MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH31634MD

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 749230-47-5
    Molecular Formula C7H4F2O3
    Formula Weight 174.1
    MDL Number MFCD07368161
    Synonym 2,3-Difluoro-6-hydroxybenzoic acid
    Chemical Name or Material 5,6-Difluorosalicylic acid

    Specifications

    Formula Weight 174.1
    MDL Number MFCD07368161
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Melting Point 181° to 184°C
    Quantity 250mg
    Molecular Formula C7H4F2O3
    CAS 749230-47-5
    Synonym 2,3-Difluoro-6-hydroxybenzoic acid
    Health Hazard 1 Warning
    Chemical Name or Material 5,6-Difluorosalicylic acid
    Assay 98%

    Description

    Solubility
    Insoluble in water.

    Notes
    Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit