Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 5,6-Dimethyl-1,10-phenanthroline, 98%
Click to view available options
Quantity:
250mg
1g
5g
Specifications
Sensitivity | Hygroscopic |
Quantity | 1g |
Melting Point | 266°C to 269°C |
Beilstein | 160918 |
Formula Weight | 208.26 |
Percent Purity | 98% |
Chemical Name or Material | 5,6-Dimethyl-1,10-phenanthroline |
Safety and Handling
EINECSNumber : 221-100-2
TSCA : Yes
Recommended Storage : Ambient temperatures