Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5,6-Dimethyl-1,10-phenanthroline, 98%

Click to view available options
Quantity:
250mg
1g
5g
3002-81-1
C14H12N2
208.264
MFCD00004983
BRPQDJPJBCQFSR-UHFFFAOYSA-N
1,10-phenanthroline, 5,6-dimethyl,acmc-1afjl,brpqdjpjbcqfsr-uhfffaoysa,5,6-dimethylpyridino 3,2-h quinoline,5,6-dimethyl-1,10-phenanthroline
76357
5,6-dimethyl-1,10-phenanthroline
CC1=C2C=CC=NC2=C3C(=C1C)C=CC=N3
3002-81-1
C14H12N2
208.264
MFCD00004983
BRPQDJPJBCQFSR-UHFFFAOYSA-N
1,10-phenanthroline, 5,6-dimethyl,acmc-1afjl,brpqdjpjbcqfsr-uhfffaoysa,5,6-dimethylpyridino 3,2-h quinoline,5,6-dimethyl-1,10-phenanthroline
76357
5,6-dimethyl-1,10-phenanthroline
CC1=C2C=CC=NC2=C3C(=C1C)C=CC=N3
Quantity 250mg
Melting Point 266°C to 269°C
Beilstein 160918
Formula Weight 208.26
Percent Purity 98%
Chemical Name or Material 5,6-Dimethyl-1,10-phenanthroline