Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Dimethyl-1H-benzotriazole Hydrate 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
4184-79-6
C8H9N3·xH2O
147.18
MFCD00149344
VVWPOQUENPOMAK-UHFFFAOYSA-N
5,6-dimethyl-1h-benzotriazole hydrate,5,6-dimethyl-1,2,3-benzotriazole hydrate,5,6-dimethyl-1h-1,2,3-benzotriazole hydrate,acmc-1aqbb,1h-benzotriazole,5,6-dimethyl,5,6-dimethylbenzotriazole, oxamethane,5,6-dimethyl-2h-benzotriazole hydrate,5,6-dimethyl-1h-benzotriazole monohydrate,5,6-dimethyl-1h-benzo d 1,2,3 triazole hydrate
16219214
5,6-dimethyl-2H-benzotriazole;hydrate
CC1=CC2=NNN=C2C=C1C.O
4184-79-6
C8H9N3·xH2O
147.18
MFCD00149344
VVWPOQUENPOMAK-UHFFFAOYSA-N
5,6-dimethyl-1h-benzotriazole hydrate,5,6-dimethyl-1,2,3-benzotriazole hydrate,5,6-dimethyl-1h-1,2,3-benzotriazole hydrate,acmc-1aqbb,1h-benzotriazole,5,6-dimethyl,5,6-dimethylbenzotriazole, oxamethane,5,6-dimethyl-2h-benzotriazole hydrate,5,6-dimethyl-1h-benzotriazole monohydrate,5,6-dimethyl-1h-benzo d 1,2,3 triazole hydrate
16219214
5,6-dimethyl-2H-benzotriazole;hydrate
CC1=CC2=NNN=C2C=C1C.O
Quantity 25g
Color Yellow to Brown
Water 11% max. (K.F.)
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 153.0°C to 156.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 147.18
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 5, 6-Dimethyl-1H-benzotriazole hydrate, 99%
Show More Show Less

EINECSNumber : 224-058-3