Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5,6-Dimethyl-1H-benzotriazole monohydrate, 99%

Click to view available options
Quantity:
5g
25g
1354973-50-4
C8H11N3O
165.196
MFCD00149344
VVWPOQUENPOMAK-UHFFFAOYSA-N
5,6-dimethyl-1h-benzotriazole hydrate,5,6-dimethyl-1,2,3-benzotriazole hydrate,5,6-dimethyl-1h-1,2,3-benzotriazole hydrate,acmc-1aqbb,1h-benzotriazole,5,6-dimethyl,5,6-dimethylbenzotriazole, oxamethane,5,6-dimethyl-2h-benzotriazole hydrate,5,6-dimethyl-1h-benzotriazole monohydrate,5,6-dimethyl-1h-benzo d 1,2,3 triazole hydrate
16219214
5,6-dimethyl-2H-benzotriazole;hydrate
CC1=CC2=NNN=C2C=C1C.O
1354973-50-4
C8H11N3O
165.196
MFCD00149344
VVWPOQUENPOMAK-UHFFFAOYSA-N
5,6-dimethyl-1h-benzotriazole hydrate,5,6-dimethyl-1,2,3-benzotriazole hydrate,5,6-dimethyl-1h-1,2,3-benzotriazole hydrate,acmc-1aqbb,1h-benzotriazole,5,6-dimethyl,5,6-dimethylbenzotriazole, oxamethane,5,6-dimethyl-2h-benzotriazole hydrate,5,6-dimethyl-1h-benzotriazole monohydrate,5,6-dimethyl-1h-benzo d 1,2,3 triazole hydrate
16219214
5,6-dimethyl-2H-benzotriazole;hydrate
CC1=CC2=NNN=C2C=C1C.O
Melting Point 153°C to 157°C
Quantity 5g
Beilstein 122933
Formula Weight 165.20 (147.18 Anhydrous)
Percent Purity 99%
Chemical Name or Material 5,6-Dimethyl-1H-benzotriazole monohydrate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 224-058-3

TSCA : No

Recommended Storage : Ambient temperatures