Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine, 99%

Click to view available options
Quantity:
1g
5g
25g
1046-56-6
C20H14N4
310.36
MFCD00006462
OTMYLOBWDNFTLO-UHFFFAOYSA-N
3-2-pyridyl-5,6-diphenyl-1,2,4-triazine,5,6-diphenyl-3-2-pyridyl-1,2,4-triazine,1,2,4-triazine, 5,6-diphenyl-3-2-pyridinyl,5,6-diphenyl-3-pyridin-2-yl-1,2,4-triazine,as-triazine, 5,6-diphenyl-3-2-pyridyl,5,6-diphenyl-3-2-pyridinyl-1,2,4-triazine,maybridge1_007490,bio7c7,acmc-1boy4,cbmicro_025706
70588
5,6-diphenyl-3-pyridin-2-yl-1,2,4-triazine
C1=CC=C(C=C1)C2=C(N=NC(=N2)C3=CC=CC=N3)C4=CC=CC=C4
1046-56-6
C20H14N4
310.36
MFCD00006462
OTMYLOBWDNFTLO-UHFFFAOYSA-N
3-2-pyridyl-5,6-diphenyl-1,2,4-triazine,5,6-diphenyl-3-2-pyridyl-1,2,4-triazine,1,2,4-triazine, 5,6-diphenyl-3-2-pyridinyl,5,6-diphenyl-3-pyridin-2-yl-1,2,4-triazine,as-triazine, 5,6-diphenyl-3-2-pyridyl,5,6-diphenyl-3-2-pyridinyl-1,2,4-triazine,maybridge1_007490,bio7c7,acmc-1boy4,cbmicro_025706
70588
5,6-diphenyl-3-pyridin-2-yl-1,2,4-triazine
C1=CC=C(C=C1)C2=C(N=NC(=N2)C3=CC=CC=N3)C4=CC=CC=C4
Applications
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine is widely utilized as a reagent for the determination of sub microgram levels of iron in serum. It is also used for the simultaneous determination of iron and copper.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.
Quantity 5g
Melting Point 189°C to 192°C
Beilstein 542151
Formula Weight 310.36
Percent Purity 99%
Chemical Name or Material 5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XY8598150

TSCA : TSCA