Learn More
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine, 99%, Thermo Scientific Chemicals
Description
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine is widely utilized as a reagent for the determination of sub microgram levels of iron in serum. It is also used for the simultaneous determination of iron and copper.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine is widely utilized as a reagent for the determination of sub microgram levels of iron in serum. It is also used for the simultaneous determination of iron and copper.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications
Melting Point | 189°C to 192°C |
Quantity | 1 g |
Beilstein | 542151 |
Solubility Information | Soluble in water. |
IUPAC Name | 5,6-diphenyl-3-pyridin-2-yl-1,2,4-triazine |
Formula Weight | 310.36 |
Percent Purity | 99% |
Chemical Name or Material | 5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 213-878-7 |
RTECSNumber | XY8598150 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.