Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (±)5,6-EET methyl ester, 0.01% w/v in ethanol

Click to view available options
Quantity:
25μg
50μg
100μg
122799-12-6
C21H34O3
334.5
MFCD00077340
XGESKIWNDBIILZ-JPFHKJGASA-N
(±)5(6)-epoxy-8Z,11Z,14Z-eicosatrienoic acid, methyl ester
13075008
methyl 4-[3-[(2Z,5Z,8Z)-tetradeca-2,5,8-trienyl]oxiran-2-yl]butanoate
CCCCCC=CCC=CCC=CCC1C(O1)CCCC(=O)OC
122799-12-6
C21H34O3
334.5
MFCD00077340
XGESKIWNDBIILZ-JPFHKJGASA-N
(±)5(6)-epoxy-8Z,11Z,14Z-eicosatrienoic acid, methyl ester
13075008
methyl 4-[3-[(2Z,5Z,8Z)-tetradeca-2,5,8-trienyl]oxiran-2-yl]butanoate
CCCCCC=CCC=CCC=CCC1C(O1)CCCC(=O)OC
UN Number UN1170
Concentration 0.01% w/v in ethanol
Formula Weight 334.5
Physical Form Liquid
Color Colorless
Quantity 50μg
Chemical Name or Material (±)5,6-EET methyl ester

GHS H Statement
H225-H371
Highly flammable liquid and vapor.
May cause damage to organs.

GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: ETHANOL SOLUTIONS

storageNote1 : -30° to -10°C