Learn More
Aat BioquestSupplier Diversity Partner 5(6)-FAM [5-(and-6)-Carboxyfluorescein] *CAS 72088-94-9* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 100
Carboxyfluorescein (commonly called FAM) and its amine-reactive succinimidyl esters are favored over FITC in bioconjugations. FAM reagents give carboxamides that are more resistant to hydrolysis.
Catalog No. 50-195-2710