Learn More
Aat BioquestSupplier Diversity Partner 5(6)-TAMRA [5(6)-Carboxytetramethylrhodamine] *CAS 98181-63-6* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 361
TAMRA is one of the most popular fluorophores used in various bioconjugations. TMR is a bright orange fluorophore. 5(6)-TAMRA is the mixture of two carboxy tetramethylrhodamine (TMR) isomers.
Catalog No. 50-195-2850