Learn More
Aat BioquestSupplier Diversity Partner 5(6)-TRITC [Tetramethylrhodamine-5-(and-6)-isothiocyanate] *CAS 95197-95-8* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 409
5(6)-TRITC is an amino-reactive labeling reagent that is widely used in preparing bioconjugates of proteins and nucleic acids. The resultant conjugates have similar spectral properties to those prepared from 5(6)-TAMRA, SE.
Catalog No. 50-195-2878