Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5,7,12,14-Pentacenetetrone, 98+%

Click to view available options
Quantity:
1g
5g
23912-79-0
C22H10O4
338.318
MFCD00075209
YZOGOBWHTVNKGA-UHFFFAOYSA-N
5,7,12,14-pentacenetetrone,2.3-phthalylanthrachinon,pentacene-5,7,12,14-tetraone,5,7,12,14-tetraoxo-5,7,12,14-tetrahydropentacene,5,7,12,14-tetrahydropentacene-5,7,12,14-tetrone
4733
pentacene-5,7,12,14-tetrone
C1=CC=C2C(=C1)C(=O)C3=CC4=C(C=C3C2=O)C(=O)C5=CC=CC=C5C4=O
23912-79-0
C22H10O4
338.318
MFCD00075209
YZOGOBWHTVNKGA-UHFFFAOYSA-N
5,7,12,14-pentacenetetrone,2.3-phthalylanthrachinon,pentacene-5,7,12,14-tetraone,5,7,12,14-tetraoxo-5,7,12,14-tetrahydropentacene,5,7,12,14-tetrahydropentacene-5,7,12,14-tetrone
4733
pentacene-5,7,12,14-tetrone
C1=CC=C2C(=C1)C(=O)C3=CC4=C(C=C3C2=O)C(=O)C5=CC=CC=C5C4=O
Quantity 1g
Formula Weight 338.32
Percent Purity ≥98%
Chemical Name or Material 5,7,12,14-Pentacenetetrone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning