Alfa Aesar™ 5,7-Dichloro-1-indanone, 97%

Manufacturer: Alfa Aesar™ H5400706

Quantity 5g

 View more versions of this product

Catalog No. AAH5400706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 448193-94-0
    Molecular Formula C9H6Cl2O
    Formula Weight 201.05
    MDL Number MFCD09908160
    Chemical Name or Material 5,7-Dichloro-1-indanone

    Specifications

    Formula Weight 201.05
    Health Hazard 1 Danger
    Molecular Formula C9H6Cl2O
    MDL Number MFCD09908160
    CAS 448193-94-0
    Health Hazard 2 GHS H Statement
    H301-H319
    Toxic if swallowed.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280-P301+P310-P305+P351+P338-P321-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 98°C
    Quantity 5g
    Chemical Name or Material 5,7-Dichloro-1-indanone
    Assay 97%

    Description

    Applications
    5,7-Dichloro-1-indanone is used as an organic chemical synthesis intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H301-H319
    Toxic if swallowed.
    Causes serious eye irritation.

    GHS P Statement
    P280-P301+P310-P305+P351+P338-P321-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit