Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,7-Dichloropyrazolo[1,5-a]pyrimidine, 95%, Thermo Scientific™

Click to view available options
Quantity:
250mg
1g
57489-77-7
C6H3Cl2N3
188.02
JMTFWCYVZOFHLR-UHFFFAOYSA-N
5,7-dichloropyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine, 5,7-dichloro, 5,7-dichloro-pyrazolo 1,5-a pyrimidine, pubchem14638, 5,7-dichloropyrazolo 1,5-a pyri-midine, 5, 7-dichloro-pyrazolo 1,5-a pyrimidine, 5,7-bis chloranyl pyrazolo 1,5-a pyrimidine
11074154
5,7-dichloropyrazolo[1,5-a]pyrimidine
C1=C2N=C(C=C(N2N=C1)Cl)Cl
57489-77-7
C6H3Cl2N3
188.02
JMTFWCYVZOFHLR-UHFFFAOYSA-N
5,7-dichloropyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine, 5,7-dichloro, 5,7-dichloro-pyrazolo 1,5-a pyrimidine, pubchem14638, 5,7-dichloropyrazolo 1,5-a pyri-midine, 5, 7-dichloro-pyrazolo 1,5-a pyrimidine, 5,7-bis chloranyl pyrazolo 1,5-a pyrimidine
11074154
5,7-dichloropyrazolo[1,5-a]pyrimidine
C1=C2N=C(C=C(N2N=C1)Cl)Cl

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point 75.0°C to 76.5°C
Assay Percent Range 94% min. (GC)
Infrared Spectrum Authentic
Quantity 250mg
Formula Weight 188.02
Percent Purity 95%
Chemical Name or Material 5,7-Dichloropyrazolo[1,5-a]pyrimidine

GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

RUO – Research Use Only