Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,7-Dihydroindolo[2,3-b]carbazole, 96%, Alfa Aesar™

Click to view available options
Quantity:
250mg
1g
111296-90-3
C18H12N2
256.308
MFCD10687177
UVHIWKCVGUMHNB-UHFFFAOYSA-N
5,7-dihydroindolo 2,3-b carbazole,5,7-dihydro-indolo 2,3-b carbazole,indolo 2,3-b carbazole,5,7-dihydro,5h,7h-indolo 2,3-b carbazole,pubchem23468,acmc-209wje,7h-indolo 2,3-b carbazole,1,3-dihydroindolo 2,3-b carbazole,indolo 2,3-arbazole, 5,7-ihydro,indolo 2,3-b carbazole, 5,7-dihydro
10332633
5,7-dihydroindolo[2,3-b]carbazole
C1=CC=C2C(=C1)C3=CC4=C(C=C3N2)NC5=CC=CC=C54
111296-90-3
C18H12N2
256.308
MFCD10687177
UVHIWKCVGUMHNB-UHFFFAOYSA-N
5,7-dihydroindolo 2,3-b carbazole,5,7-dihydro-indolo 2,3-b carbazole,indolo 2,3-b carbazole,5,7-dihydro,5h,7h-indolo 2,3-b carbazole,pubchem23468,acmc-209wje,7h-indolo 2,3-b carbazole,1,3-dihydroindolo 2,3-b carbazole,indolo 2,3-arbazole, 5,7-ihydro,indolo 2,3-b carbazole, 5,7-dihydro
10332633
5,7-dihydroindolo[2,3-b]carbazole
C1=CC=C2C(=C1)C3=CC4=C(C=C3N2)NC5=CC=CC=C54
Quantity 1g
Density 1.40g/mL
Melting Point 348°C to 360°C
Color Brown
Formula Weight 256.3
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 5,7-Dihydroindolo[2,3-b]carbazole