Alfa Aesar™ 5,7-Dimethoxycoumarin, 98%

Manufacturer: Alfa Aesar™ B2322606

Quantity 5g

 View more versions of this product

Catalog No. AAB2322606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 487-06-9
    Molecular Formula C11H10O4
    Formula Weight 206.2
    MDL Number MFCD00006870
    Merck Index 14,5492
    Synonym Citropten; Limettin
    Chemical Name or Material 5,7-Dimethoxycoumarin

    Specifications

    Formula Weight 206.2
    Melting Point 146° to 149°C
    MDL Number MFCD00006870
    Quantity 5g
    Merck Index 14,5492
    Synonym Citropten; Limettin
    Molecular Formula C11H10O4
    CAS 487-06-9
    Chemical Name or Material 5,7-Dimethoxycoumarin
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit