Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,7-Dimethylisatin, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
39603-24-2
C10H9NO2
175.187
MFCD00047219
HFZSCCJTJGWTDZ-UHFFFAOYSA-N
5,7-dimethylisatin,5,7-dimethylindoline-2,3-dione,5,7-dimethyl-indole-2,3-dione,5,7-dimethyl-2,3-dihydro-1h-indole-2,3-dione,1h-indole-2,3-dione, 5,7-dimethyl,indole-2,3-dione, 5,7-dimethyl,1h-indole-2,3-dione, 5,7-dimethyl-9ci,acmc-20a4zq,isatin-based compound, 48,1h-indole-2,3-dione,5,7-dimethyl
38296
5,7-dimethyl-1H-indole-2,3-dione
CC1=CC(=C2C(=C1)C(=O)C(=O)N2)C
39603-24-2
C10H9NO2
175.187
MFCD00047219
HFZSCCJTJGWTDZ-UHFFFAOYSA-N
5,7-dimethylisatin,5,7-dimethylindoline-2,3-dione,5,7-dimethyl-indole-2,3-dione,5,7-dimethyl-2,3-dihydro-1h-indole-2,3-dione,1h-indole-2,3-dione, 5,7-dimethyl,indole-2,3-dione, 5,7-dimethyl,1h-indole-2,3-dione, 5,7-dimethyl-9ci,acmc-20a4zq,isatin-based compound, 48,1h-indole-2,3-dione,5,7-dimethyl
38296
5,7-dimethyl-1H-indole-2,3-dione
CC1=CC(=C2C(=C1)C(=O)C(=O)N2)C
Quantity 1g
Color Undesignated
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 175.19
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material 5, 7-Dimethylisatin, 95%
Show More Show Less

RTECSNumber : NL7930000