Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Acetyl-2-norbornene 98.0+%, TCI America™

Click to view available options
Quantity:
25g
5063-03-6
C9H12O
136.19
MFCD00167570
NIMLCWCLVJRPFY-UHFFFAOYNA-N
2-acetyl-5-norbornene,5-acetyl-2-norbornene,1-bicyclo 2.2.1 hept-5-en-2-yl ethanone,5-acetylbicyclo 2.2.1 hept-2-ene,methyl 5-norbornen-2-yl ketone,1-5-bicyclo 2.2.1 hept-2-enyl ethanone,ethanone, 1-bicyclo 2.2.1 hept-5-en-2-yl,1-bicyclo 2.2.1 hept-5-en-2-yl ethan-1-one,5-acetylnorbornene
107366
1-(5-bicyclo[2.2.1]hept-2-enyl)ethanone
CC(=O)C1CC2CC1C=C2
5063-03-6
C9H12O
136.19
MFCD00167570
NIMLCWCLVJRPFY-UHFFFAOYNA-N
2-acetyl-5-norbornene,5-acetyl-2-norbornene,1-bicyclo 2.2.1 hept-5-en-2-yl ethanone,5-acetylbicyclo 2.2.1 hept-2-ene,methyl 5-norbornen-2-yl ketone,1-5-bicyclo 2.2.1 hept-2-enyl ethanone,ethanone, 1-bicyclo 2.2.1 hept-5-en-2-yl,1-bicyclo 2.2.1 hept-5-en-2-yl ethan-1-one,5-acetylnorbornene
107366
1-(5-bicyclo[2.2.1]hept-2-enyl)ethanone
CC(=O)C1CC2CC1C=C2
Color Yellow
Boiling Point 82°C
Quantity 25g
Formula Weight 136.19
Physical Form Clear Liquid at 20°C
Percent Purity ≥95.0% (GC)
Chemical Name or Material 5-Acetyl-2-norbornene