Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Acetyl-2-thiopheneboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
206551-43-1
C6H7BO3S
170
MFCD01075681
DCNMATSPQKWETQ-UHFFFAOYSA-N
5-acetylthiophene-2-boronic acid,5-acetyl-2-thiopheneboronic acid,5-acetyl-2-thienylboronic acid,5-acetylthiophen-2-yl boronic acid,5-acetyl-2-thienyl boronic acid,5-acetylthiophen-2-boronic acid,boronic acid, 5-acetyl-2-thienyl,2-acetylthien-5-yl boronic acid,5-acetylthien-2-yl boronic acid,1-5-dihydroxyboranyl thiophen-2-yl ethan-1-one
4143377
(5-acetylthiophen-2-yl)boronic acid
B(C1=CC=C(S1)C(=O)C)(O)O
206551-43-1
C6H7BO3S
170
MFCD01075681
DCNMATSPQKWETQ-UHFFFAOYSA-N
5-acetylthiophene-2-boronic acid,5-acetyl-2-thiopheneboronic acid,5-acetyl-2-thienylboronic acid,5-acetylthiophen-2-yl boronic acid,5-acetyl-2-thienyl boronic acid,5-acetylthiophen-2-boronic acid,boronic acid, 5-acetyl-2-thienyl,2-acetylthien-5-yl boronic acid,5-acetylthien-2-yl boronic acid,1-5-dihydroxyboranyl thiophen-2-yl ethan-1-one
4143377
(5-acetylthiophen-2-yl)boronic acid
B(C1=CC=C(S1)C(=O)C)(O)O
Packaging Glass bottle
Infrared Spectrum Authentic
Color White
Quantity 5g
Melting Point 148.0°C to 150.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Solubility Information Solubility in water: ca 2.5%
Formula Weight 170
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 5-Acetyl-2-thiopheneboronic acid
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning