Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Acetylsalicylamide, 98%

Click to view available options
Quantity:
5g
25g
100g
40187-51-7
C9H9NO3
179.175
MFCD00049222
LWAQTCWTCCNHJR-UHFFFAOYSA-N
5-acetylsalicylamide,benzamide, 5-acetyl-2-hydroxy,5-acetyl-2-hydroxy-benzamide,5-acetyl salicylamide,acmc-1alb7,dsstox_cid_31525,dsstox_rid_97410,dsstox_gsid_57736,4-10-00-03635 beilstein handbook reference
198212
5-acetyl-2-hydroxybenzamide
CC(=O)C1=CC(=C(C=C1)O)C(=O)N
40187-51-7
C9H9NO3
179.175
MFCD00049222
LWAQTCWTCCNHJR-UHFFFAOYSA-N
5-acetylsalicylamide,benzamide, 5-acetyl-2-hydroxy,5-acetyl-2-hydroxy-benzamide,5-acetyl salicylamide,acmc-1alb7,dsstox_cid_31525,dsstox_rid_97410,dsstox_gsid_57736,4-10-00-03635 beilstein handbook reference
198212
5-acetyl-2-hydroxybenzamide
CC(=O)C1=CC(=C(C=C1)O)C(=O)N
Quantity 100g
Melting Point 220°C to 222°C
Formula Weight 179.18
Percent Purity 98%
Chemical Name or Material 5-Acetylsalicylamide

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 254-830-5

RTECSNumber : CU8702280

TSCA : No

Recommended Storage : Ambient temperatures