Alfa Aesar™ 5-Acetyluracil, 98%

Manufacturer: Alfa Aesar™ 4437803

Quantity 1g

 View more versions of this product

Catalog No. AA4437803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6214-65-9
    Molecular Formula C6H6N2O3
    Formula Weight 154.11
    MDL Number MFCD00151968
    Synonym 5-Acetyl-1H-pyrimidine-2,4-dione
    Chemical Name or Material 5-Acetyluracil

    Specifications

    Formula Weight 154.11
    Physical Form Powder
    MDL Number MFCD00151968
    Molecular Formula C6H6N2O3
    Quantity 1g
    CAS 6214-65-9
    Synonym 5-Acetyl-1H-pyrimidine-2,4-dione
    Chemical Name or Material 5-Acetyluracil
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit