Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Amino-1-(2-hydroxyethyl)-1H-pyrazole-4-carbonitrile, 97%

Click to view available options
Quantity:
250mg
1g
5g
5346-53-2
C6H8N4O
152.157
MFCD00020739
FOZZJAVSCUBGPM-UHFFFAOYSA-N
5-amino-1-2-hydroxyethyl-1h-pyrazole-4-carbonitrile,5-amino-4-cyano-1-2-hydroxyethyl-pyrazole,5-amino-1-2-hydroxyethyl pyrazole-4-carbonitrile,5-amino-4-cyano-1-2-hydroxyethyl pyrazole,4-cyano-1-2-hydroxyethyl-1h-pyrazol-5-amine,5-amino-4-cyano-1-2-hydroethyl pyrazole,5-amino-1-2-hydroxy-ethyl-1h-pyrazole-4-carbonitrile,1h-pyrazole-4-carbonitrile, 5-amino-1-2-hydroxyethyl,1-,a-hydroxyethyl-4-cyano-5-aminopyrazole,zlchem 605
219852
5-amino-1-(2-hydroxyethyl)pyrazole-4-carbonitrile
C1=NN(C(=C1C#N)N)CCO
5346-53-2
C6H8N4O
152.157
MFCD00020739
FOZZJAVSCUBGPM-UHFFFAOYSA-N
5-amino-1-2-hydroxyethyl-1h-pyrazole-4-carbonitrile,5-amino-4-cyano-1-2-hydroxyethyl-pyrazole,5-amino-1-2-hydroxyethyl pyrazole-4-carbonitrile,5-amino-4-cyano-1-2-hydroxyethyl pyrazole,4-cyano-1-2-hydroxyethyl-1h-pyrazol-5-amine,5-amino-4-cyano-1-2-hydroethyl pyrazole,5-amino-1-2-hydroxy-ethyl-1h-pyrazole-4-carbonitrile,1h-pyrazole-4-carbonitrile, 5-amino-1-2-hydroxyethyl,1-,a-hydroxyethyl-4-cyano-5-aminopyrazole,zlchem 605
219852
5-amino-1-(2-hydroxyethyl)pyrazole-4-carbonitrile
C1=NN(C(=C1C#N)N)CCO
Color White
Quantity 5g
UN Number UN3439
Beilstein 155176
Formula Weight 152.16
Physical Form Crystalline Solid
Percent Purity 97%
Chemical Name or Material 5-Amino-1-(2-hydroxyethyl)-1H-pyrazole-4-carbonitrile

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

storageNote1 : -30° to -10°C