Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Amino-1-indanone, 95%

Click to view available options
Quantity:
250mg
1g
3470-54-0
C9H9NO
147.177
MFCD00099466
HODOSJNSRPXYBH-UHFFFAOYSA-N
5-aminoindan-1-one,5-amino-1-indanone,5-amino-2,3-dihydro-1h-inden-1-one,1h-inden-1-one, 5-amino-2,3-dihydro,5-aminoindanone,5-amino-1-oxoindane,5-amino-indan-1-one,pubchem14476,5-amino-1-indanone #,5-azanyl-2,3-dihydroinden-1-one
312890
5-amino-2,3-dihydroinden-1-one
C1CC(=O)C2=C1C=C(C=C2)N
3470-54-0
C9H9NO
147.177
MFCD00099466
HODOSJNSRPXYBH-UHFFFAOYSA-N
5-aminoindan-1-one,5-amino-1-indanone,5-amino-2,3-dihydro-1h-inden-1-one,1h-inden-1-one, 5-amino-2,3-dihydro,5-aminoindanone,5-amino-1-oxoindane,5-amino-indan-1-one,pubchem14476,5-amino-1-indanone #,5-azanyl-2,3-dihydroinden-1-one
312890
5-amino-2,3-dihydroinden-1-one
C1CC(=O)C2=C1C=C(C=C2)N
Melting Point 184°C to 186°C
Quantity 250mg
UN Number UN2811
Beilstein 1636158
Formula Weight 147.18
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 5-Amino-1-indanone

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.