Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-1-methyl-1H-pyrazole, 97%

Catalog No. p-7077386
Click to view available options
Quantity:
25 g
100 g

CAS: 1192-21-8 | C4H7N3 | 97.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1192-21-8
Molecular Formula C4H7N3
Molecular Weight (g/mol) 97.12
InChI Key JESRNIJXVIFVOV-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazol-5-amine, 1-methyl-1h-pyrazol-5-ylamine, 1-methyl-5-aminopyrazole, 5-amino-1-methylpyrazole, 5-amino-1-methyl-1h-pyrazole, 1h-pyrazol-5-amine, 1-methyl, 2-methyl-2h-pyrazol-3-ylamine, 5-amino-1-methyl-pyrazole, 2-methyl-2h-pyrazol-3-amine, 1-methylpyrazole-5-ylamine
PubChem CID 136927
IUPAC Name 2-methylpyrazol-3-amine
SMILES CN1C(=CC=N1)N

Specifications

Melting Point 73.0°C to 76.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 25 g
Formula Weight 97.12
Percent Purity 97%
Chemical Name or Material 5-Amino-1-methyl-1H-pyrazole
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Take off contaminated clothing and wash before reuse.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.