Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-1-pentanol, 50 wt.% aqueous solution, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
800mL
Packaging:
Glass bottle
2508-29-4,7732-18-5
C5H13NO
103.16
MFCD00008237
LQGKDMHENBFVRC-UHFFFAOYSA-N
5-amino-1-pentanol,5-aminopentanol,1-pentanol, 5-amino,pentanol, 5-amino,5-amino-1-pentanol in water,5-amino pentanol,5-amino-l-pentanol,5-aminopentanol-1,5-hydroxypentylamine,1-amino-5-pentanol
75634
5-aminopentan-1-ol
C(CCN)CCO
2508-29-4,7732-18-5
C5H13NO
103.16
MFCD00008237
LQGKDMHENBFVRC-UHFFFAOYSA-N
5-amino-1-pentanol,5-aminopentanol,1-pentanol, 5-amino,pentanol, 5-amino,5-amino-1-pentanol in water,5-amino pentanol,5-amino-l-pentanol,5-aminopentanol-1,5-hydroxypentylamine,1-amino-5-pentanol
75634
5-aminopentan-1-ol
C(CCN)CCO
Specific Gravity 0.987
Appearance Clear and colorless
Quantity 25mL
Infrared Spectrum Authentic
Density 0.9870g/mL
Packaging Glass bottle
Flash Point >100°C
CAS Min % 48.0,48.0
CAS Max % 52.0,52.0
Linear Formula OH(CH2)5NH2
Solubility Information Solubility in water: miscible.
Formula Weight 103.16
Percent Purity 48.0 to 52.0%
Chemical Name or Material 5-Amino-1-pentanol
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 219-718-2