Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-1H-benzimidazole-2-thiol, 97%, Maybridge

Click to view available options
Quantity:
1g
10g
50g
2818-66-8
C7H7N3S
165.214
BXDMTLVCACMNJO-UHFFFAOYSA-N
5-amino-2-mercaptobenzimidazole,5-amino-2-benzimidazolethiol,5-amino-1h-benzo d imidazole-2-thiol,5-amino-1h-benzimidazole-2-thiol,5-aminobenzimidazole-2-thiol,2h-benzimidazole-2-thione, 5-amino-1,3-dihydro,2-benzimidazolethiol, 5-amino,5-amino-1h-1,3-benzodiazole-2-thiol,2-mercapto-5-amino benzimidazole,5-amino-3h-1,3-benzodiazole-2-thiol
2734002
5-amino-1,3-dihydrobenzimidazole-2-thione
C1=CC2=C(C=C1N)NC(=S)N2
2818-66-8
C7H7N3S
165.214
BXDMTLVCACMNJO-UHFFFAOYSA-N
5-amino-2-mercaptobenzimidazole,5-amino-2-benzimidazolethiol,5-amino-1h-benzo d imidazole-2-thiol,5-amino-1h-benzimidazole-2-thiol,5-aminobenzimidazole-2-thiol,2h-benzimidazole-2-thione, 5-amino-1,3-dihydro,2-benzimidazolethiol, 5-amino,5-amino-1h-1,3-benzodiazole-2-thiol,2-mercapto-5-amino benzimidazole,5-amino-3h-1,3-benzodiazole-2-thiol
2734002
5-amino-1,3-dihydrobenzimidazole-2-thione
C1=CC2=C(C=C1N)NC(=S)N2
Quantity 50g
Assay Percent Range 97%
Formula Weight 165.22
Chemical Name or Material 5-Amino-1H-benzimidazole-2-thiol