Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-Amino-1H-tetrazole monohydrate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
5-Aminotetrazole monohydrate is a useful reagent in condensation reactions to form pyrimidines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Aminotetrazole monohydrate is a useful reagent in condensation reactions to form pyrimidines.
Solubility
Soluble in water, ethanol (1.36%).
Notes
Store away from strong oxidizing agents, strong acids, acid chlorides, acid anhydrides.
5-Aminotetrazole monohydrate is a useful reagent in condensation reactions to form pyrimidines.
Solubility
Soluble in water, ethanol (1.36%).
Notes
Store away from strong oxidizing agents, strong acids, acid chlorides, acid anhydrides.
Specifications
Melting Point | ∼203°C (decomposition) |
pH | 3.7 |
Quantity | 500 g |
UN Number | UN1325 |
Beilstein | 108872 |
Solubility Information | Soluble in water,ethanol (1.36%). |
Formula Weight | 103.08 (85.06 Anhydrous) |
Percent Purity | 99% |
Chemical Name or Material | 5-Amino-1H-tetrazole monohydrate |
Safety and Handling
Hazard Category | H302-H315-H318-H335 |
Hazard Statement | GHS H Statement H240-H228-H315-H319-H335 Heating may cause an explosion. Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S. |
EINECSNumber | 224-581-7 |
RTECSNumber | XF7465000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction