Alfa Aesar™ 5-Amino-2-(2,4-dichlorophenoxy)pyridine, 99%

Manufacturer: Alfa Aesar™ H5068303

Quantity 1g

 View more versions of this product

Catalog No. AAH5068303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 85686-48-2
    Molecular Formula C11H8Cl2N2O
    Formula Weight 255.11
    MDL Number MFCD00052643
    Synonym 2,4-Dichlorophenoxy-5-pyridinamine
    Chemical Name or Material 5-Amino-2-(2,4-dichlorophenoxy)pyridine

    Specifications

    Formula Weight 255.11
    Quantity 1g
    Molecular Formula C11H8Cl2N2O
    MDL Number MFCD00052643
    Synonym 2,4-Dichlorophenoxy-5-pyridinamine
    CAS 85686-48-2
    Chemical Name or Material 5-Amino-2-(2,4-dichlorophenoxy)pyridine
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit