Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Amino-2,2-difluoro-1,3-benzodioxole, 97+%

Click to view available options
Quantity:
1g
5g
1544-85-0
C7H5F2NO2
173.119
MFCD00190144
CVYQRDKVWVBOFP-UHFFFAOYSA-N
2,2-difluoro-5-aminobenzodioxole,2,2-difluorobenzo d 1,3 dioxol-5-amine,2,2-difluoro-5-amino-1,3-benzodioxole,5-amino-2,2-difluorobenzodioxole,5-amino-2,2-difluoro-1,3-benzodioxole,2,2-difluoro-2h-1,3-benzodioxol-5-amine,1,3-benzodioxol-5-amine, 2,2-difluoro,3,4-difluoromethylenedioxy aniline,3,4-difluoromethylene dioxy aniline,2,2-difluoro-benzo 1,3 dioxol-5-ylamine
2736893
2,2-difluoro-1,3-benzodioxol-5-amine
C1=CC2=C(C=C1N)OC(O2)(F)F
1544-85-0
C7H5F2NO2
173.119
MFCD00190144
CVYQRDKVWVBOFP-UHFFFAOYSA-N
2,2-difluoro-5-aminobenzodioxole,2,2-difluorobenzo d 1,3 dioxol-5-amine,2,2-difluoro-5-amino-1,3-benzodioxole,5-amino-2,2-difluorobenzodioxole,5-amino-2,2-difluoro-1,3-benzodioxole,2,2-difluoro-2h-1,3-benzodioxol-5-amine,1,3-benzodioxol-5-amine, 2,2-difluoro,3,4-difluoromethylenedioxy aniline,3,4-difluoromethylene dioxy aniline,2,2-difluoro-benzo 1,3 dioxol-5-ylamine
2736893
2,2-difluoro-1,3-benzodioxol-5-amine
C1=CC2=C(C=C1N)OC(O2)(F)F
Applications
5-Amino-2,2-difluoro-1,3-benzodioxole is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Light sensitive. Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Odor Recognizable
Quantity 5g
Boiling Point 95°C to 97°C (12mmHg)
UN Number UN2810
Beilstein 1343593
Formula Weight 173.12
Percent Purity ≥97%
Chemical Name or Material 5-Amino-2,2-difluoro-1,3-benzodioxole

GHS H Statement
H301-H312-H332-H315-H319
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.