Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Amino-2,4,6-triiodoisophthalic acid, 95%

Manufacturer:  Alfa Aesar™ B2217830

Quantity 250g

 View more versions of this product

Catalog No. AAB2217830


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

35453-19-1
558.84
5-Amino-2,4,6-triiodobenzene-1,3-dicarboxylic acid
C8H4I3NO4
MFCD00190167
5-Amino-2,4,6-triiodoisophthalic acid

Specifications

35453-19-1
558.84
MFCD00190167
5-Amino-2,4,6-triiodobenzene-1,3-dicarboxylic acid
95%
C8H4I3NO4
265° to 270°C
250g
5-Amino-2,4,6-triiodoisophthalic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit