Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Amino-2,4,6-triiodoisophthalic acid, 95%

Click to view available options
Quantity:
10g
50g
250g
35453-19-1
C8H4I3NO4
558.836
MFCD00190167
JEZJSNULLBSYHV-UHFFFAOYSA-N
5-amino-2,4,6-triiodoisophthalic acid,unii-a17g990rt0,5-amino-2,4,6-triiodo-1,3-benzenedicarboxylic acid,5-amino-2,4,6-iodoisophthalic acid,1,3-benzenedicarboxylic acid, 5-amino-2,4,6-triiodo,5-amino-2,4,6-triiodoisophthalicacid,isophthalic acid, 5-amino-2,4,6-triiodo,acmc-209ih0,ksc224s6b,aronis25361
3015783
5-amino-2,4,6-triiodobenzene-1,3-dicarboxylic acid
C1(=C(C(=C(C(=C1I)N)I)C(=O)O)I)C(=O)O
35453-19-1
C8H4I3NO4
558.836
MFCD00190167
JEZJSNULLBSYHV-UHFFFAOYSA-N
5-amino-2,4,6-triiodoisophthalic acid,unii-a17g990rt0,5-amino-2,4,6-triiodo-1,3-benzenedicarboxylic acid,5-amino-2,4,6-iodoisophthalic acid,1,3-benzenedicarboxylic acid, 5-amino-2,4,6-triiodo,5-amino-2,4,6-triiodoisophthalicacid,isophthalic acid, 5-amino-2,4,6-triiodo,acmc-209ih0,ksc224s6b,aronis25361
3015783
5-amino-2,4,6-triiodobenzene-1,3-dicarboxylic acid
C1(=C(C(=C(C(=C1I)N)I)C(=O)O)I)C(=O)O
Melting Point 265°C to 270°C
Quantity 10g
Formula Weight 558.84
Percent Purity 95%
Chemical Name or Material 5-Amino-2,4,6-triiodoisophthalic acid

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger