Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Amino-2,4-dichloropyridine, 97%

Click to view available options
Quantity:
250mg
1g
7321-93-9
C5H4Cl2N2
163.001
MFCD00234054
FBGVTWONYOCYGA-UHFFFAOYSA-N
5-amino-2,4-dichloropyridine,4,6-dichloro-pyridin-3-ylamine,3-pyridinamine, 4,6-dichloro,4,6-dichloro-3-pyridinamine,4,6-dichloropyridin-3-ylamine,pubchem17045,acmc-209opx,2,4-dichloro-5-aminopyridine,4,6-dichloropyridine-3-amine,3-pyridinamine,4,6-dichloro
595729
4,6-dichloropyridin-3-amine
C1=C(C(=CN=C1Cl)N)Cl
7321-93-9
C5H4Cl2N2
163.001
MFCD00234054
FBGVTWONYOCYGA-UHFFFAOYSA-N
5-amino-2,4-dichloropyridine,4,6-dichloro-pyridin-3-ylamine,3-pyridinamine, 4,6-dichloro,4,6-dichloro-3-pyridinamine,4,6-dichloropyridin-3-ylamine,pubchem17045,acmc-209opx,2,4-dichloro-5-aminopyridine,4,6-dichloropyridine-3-amine,3-pyridinamine,4,6-dichloro
595729
4,6-dichloropyridin-3-amine
C1=C(C(=CN=C1Cl)N)Cl
Quantity 250mg
Color Brown
UN Number UN2811
Beilstein 119452
Formula Weight 163
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 5-Amino-2,4-dichloropyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

storageNote1 : -30° to -10°C