Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Amino-2-bromo-3-methylpyridine, 98%

Click to view available options
Quantity:
5g
25g
38186-83-3
C6H7BrN2
187.04
MFCD03095085
IEONGQZSAUAAKH-UHFFFAOYSA-N
5-amino-2-bromo-3-picoline,5-amino-2-bromo-3-methylpyridine,3-pyridinamine, 6-bromo-5-methyl,6-bromo-5-methyl-3-pyridylamine,6-bromo-5-methyl-3-aminopyridine,pubchem1129,ksc985c1p,2-bromo-3-methyl-5-aminopyridine,2-bromo-5-amino-3-methylpyridine,6-bromo-5-methyl-pyridin-3-ylamine
12362446
6-bromo-5-methylpyridin-3-amine
CC1=CC(=CN=C1Br)N
38186-83-3
C6H7BrN2
187.04
MFCD03095085
IEONGQZSAUAAKH-UHFFFAOYSA-N
5-amino-2-bromo-3-picoline,5-amino-2-bromo-3-methylpyridine,3-pyridinamine, 6-bromo-5-methyl,6-bromo-5-methyl-3-pyridylamine,6-bromo-5-methyl-3-aminopyridine,pubchem1129,ksc985c1p,2-bromo-3-methyl-5-aminopyridine,2-bromo-5-amino-3-methylpyridine,6-bromo-5-methyl-pyridin-3-ylamine
12362446
6-bromo-5-methylpyridin-3-amine
CC1=CC(=CN=C1Br)N
Quantity 25g
Melting Point 97°C to 100°C
Color Gray
UN Number UN2811
Beilstein 471832
Formula Weight 187.04
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 5-Amino-2-bromo-3-methylpyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

storageNote1 : 2° to 8°C