Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-bromo-6-methylpyridine, 98%, Thermo Scientific™

Catalog Number AAH6475614
Click to view available options
Quantity:
1 g
5 g
25 g
126325-47-1
C6H7BrN2
187.04
MFCD03095086
UBTQJSZELCWGCN-UHFFFAOYSA-N
5-amino-2-bromo-6-picoline, 3-amino-6-bromo-2-methylpyridine, 2-bromo-5-amino-6-methylpyridine, 3-amino-6-bromo-2-picoline, 5-amino-2-bromo-6-methylpyridine, 2-bromo-5-amino-6-picoline, 6-bromo-2-methyl-3-pyridylamine, 6-bromo-2-methyl-pyridin-3-ylamine, 2-methyl-3-amino-6-bromopyridine, pubchem1117
14418045
6-bromo-2-methylpyridin-3-amine
CC1=C(N)C=CC(Br)=N1
126325-47-1
C6H7BrN2
187.04
MFCD03095086
UBTQJSZELCWGCN-UHFFFAOYSA-N
5-amino-2-bromo-6-picoline, 3-amino-6-bromo-2-methylpyridine, 2-bromo-5-amino-6-methylpyridine, 3-amino-6-bromo-2-picoline, 5-amino-2-bromo-6-methylpyridine, 2-bromo-5-amino-6-picoline, 6-bromo-2-methyl-3-pyridylamine, 6-bromo-2-methyl-pyridin-3-ylamine, 2-methyl-3-amino-6-bromopyridine, pubchem1117
14418045
6-bromo-2-methylpyridin-3-amine
CC1=C(N)C=CC(Br)=N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 110°C to 111°C
Color White
Quantity 25 g
Beilstein 6190039
Formula Weight 187.04
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Amino-2-bromo-6-methylpyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit